missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ FcXP Pre-packed Column

Product Code. 18406428 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 x 5 mL Column
5 x 1 mL Columns
Unit Size:
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406428 1 x 5 mL Column 5mL
18486468 5 x 1 mL Columns 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18406428 Supplier Thermo Scientific™ Supplier No. 494371205

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ CaptureSelect™ FcXP Affinity Matrix is offered here in a pre-packed format with connectors that are compatible with standard chromatography equipment.

CaptureSelect FcXP Affinity Matrix is offered here in a pre-packed format with connectors that are compatible with standard chromatography equipment.

CaptureSelect FcXP Affinity Matrix is the successor to CaptureSelect FcXL Affinity Matrix, specifically designed for higher binding capacity. The selectivity is identical; the ligand recognizes the CH3 domain of all human IgG subclasses (IgG1, 2, 3, and 4). But the matrix features an increased binding capacity (>40 g IgG/L), while maintaining the elution characteristics of CaptureSelect FcXL Affinity Matrix.

Features of this affinity matrix include:

  • Mild elution, making it suitable for Fc fusion proteins
  • Human specific, no binding to bovine antibodies
  • Excellent scalability
  • Non-animal derived

CaptureSelect FcXP pre-packed columns can be used for the purification of IgG and Fc fusion proteins directly from complex source materials and enable a robust and efficient purification process with excellent purity in one step.

Characteristics:

  • Matrix: agarose-based, epoxide activated
  • Average particle size: 65 ± 10 μm
  • Ligand: CaptureSelect FcXP affinity ligand
  • Ligand coupling method: epoxide coupling
  • Binding capacity: 40–50 g IgG/L resin depending on flow rate, column height, and contact time
  • Elution conditions: 20 mM citric acid or acetic acid, pH 3–4, or 100 mM Tris, 2.0 M MgCl2, 40% (v/v) propylene glycol, pH 7
  • Flow characteristics: 150–300 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol

Specifications

Binding Capacity 40 to 50 g IgG/L resin
Flow Rate 150 to 300 cm/hr. (Up to 2 bar)
Matrix Agarose-based, epoxide-activated
Content And Storage Store at 2°C to 8°C.
Shipping Condition Wet Ice
Particle Size 65 ±10 μm
Product Line CaptureSelect
Quantity 1 x 5 mL Column
Type Pre-packed Column
Ligand Type FcXP affinity ligand

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.