Learn More
Thermo Scientific™ CaptureSelect™ FcXP Ligand Leakage ELISA
Shop All Thermo Scientific ProductsDescription
CaptureSelect FcXP Ligand Leakage ELISA is designed to detect possible leached ligand of antibodies or antibody fragments purified using CaptureSelect FcXP Affinity Matrix. The FcXP affinity ligand is a 13-kD recombinant protein, expressed in yeast and covalently coupled to a chromatography matrix for high chemical stability and low leakage. During the chromatography process small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the protein.
This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect FcXP Ligand Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.
Specifications
Specifications
| Content And Storage | 3 x 0.1 mL, store at -20°C. |
| Shipping Condition | Dry Ice |
| Form | Reagent Solution |
| Product Line | CaptureSelect |
| Quantity | 1 assay x 96-well plate |
| Target | CaptureSelect FcXP Ligand |
| Type | ELISA Kit |
| Sufficient For | 1 x 96-well plate |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.