missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ Factor X Ligand Leakage ELISA

Product Code. 18486748 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 assay x 96-well plate
10 assay x 96-well plate
Unit Size:
Each
Pack of 10
This item is not returnable. View return policy

Product Code. 18486748

Brand: Thermo Scientific™ 810370210

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ CaptureSelect™ Factor X Ligand Leakage ELISA is designed to detect possible leached ligand of Factor X purified using CaptureSelect Factor X Affinity Matrix.

The Factor X affinity ligand is a 13-kD recombinant protein expressed in yeast and covalently coupled to a chromatography matrix to give high chemical stability and low leakage. During the chromatography process small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the protein. This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL.

The CaptureSelect Factor X Ligand Leakage ELISA is designed to minimize interference and to provide accurate quantitation even in the presence of the protein of interest. It can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.

Specifications

Content And Storage 3 x 1 mL, store at -20°C.
Shipping Condition Dry Ice
Form Reagent Solution
Product Line CaptureSelect
Quantity 10 assay x 96-well plate
Target CaptureSelect Factor X Ligand
Type ELISA Kit
Sufficient For 10 x 96-well plates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.