Learn More
Thermo Scientific™ CaptureSelect™ Factor X Ligand Leakage ELISA
Shop All Thermo Scientific ProductsDescription
The Factor X affinity ligand is a 13-kD recombinant protein expressed in yeast and covalently coupled to a chromatography matrix to give high chemical stability and low leakage. During the chromatography process small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the protein. This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL.
The CaptureSelect Factor X Ligand Leakage ELISA is designed to minimize interference and to provide accurate quantitation even in the presence of the protein of interest. It can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.
Specifications
Specifications
| Content And Storage | 3 x 0.1 mL, store at -20°C. |
| Shipping Condition | Dry Ice |
| Form | Reagent Solution |
| Product Line | CaptureSelect |
| Quantity | 1 assay x 96-well plate |
| Target | CaptureSelect Factor X Ligand |
| Type | ELISA Kit |
| Sufficient For | 1 x 96-well plate |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.