missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ CH1-XL Pre-packed Column

Product Code. 18466568 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 x 5 mL Column
5 x 1 mL Columns
Unit Size:
Each
Pack of 5
This item is not returnable. View return policy

Product Code. 18466568

Brand: Thermo Scientific™ 494346201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ CaptureSelect™ CH1-XL Affinity Matrix is an improved version of CaptureSelect IgG-CH1 Affinity Matrix, featuring a wider elution range (pH 3 to 4.5), increased binding capacity, and improved stability at higher pH, offered here in pre-packed format.

CaptureSelect CH1-XL Affinity Matrix is an improved version of CaptureSelect IgG-CH1 Affinity Matrix, featuring a wider elution range (pH 3.0–4.5), increased binding capacity, and improved stability at higher pH, offered here in pre-packed format.

CaptureSelect CH1-XL Affinity Matrix recognizes the CH1 domain of human IgG antibodies, enabling purification of all subclasses (1, 2, 3, and 4) of human IgG and Fab fragments, independent of the light-chain iso-type and independent of the source material. Due to its unique selectivity for the CH1 domain, no co-purification of free light-chain contaminants will occur. Since no cross-binding is observed for bovine IgG, the product can be used for the purification of IgGs and antibody fragments in the presence of fetal calf serum.

The affinity matrix is offered in a pre-packed format and consists of 5 x 1 mL, or 1 x 5 mL columns with connectors, that are compatible with standard chromatography equipment.

Specifications

Content And Storage Store in refrigerator (2°C to 8°C).
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Product Line CaptureSelect
Quantity 5 x 1 mL Columns
Type Pre-packed Column
Ligand Type CH1-XL ligand
Stationary Phase CH1-XL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.