missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ Bovine IgA Affinity Matrix

Product Code. 18496658 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 mL
10 mL
Unit Size:
10mL
5mL
Product Code. Quantity unitSize
18496658 5 mL 5mL
18456408 10 mL 10mL
2 options
This item is not returnable. View return policy

Product Code. 18496658

Brand: Thermo Scientific™ 2943511005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ CaptureSelect™ Bovine IgA Affinity Matrix has been designed specifically for the purification of bovine IgA from whey.

CaptureSelect Bovine IgA Affinity Matrix has been designed specifically for the purification of bovine IgA from whey.

Features of this affinity matrix include:

  • Binds monomeric, dimeric, and secretory bovine IgA
  • One-step purification from whey
  • Excellent scalability

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: agarose-based, aldehyde-activated
  • Average particle size: 65 ± 10 μm
  • Ligand: CaptureSelect Bovine IgA affinity ligand
  • Ligand coupling method: aldehyde coupling via NH2 residues of the ligand
  • Binding capacity: ∼10 g/L resin
  • Elution conditions: 0.1 M citric acid pH 3.0 or 0.1M glycine pH 3.0
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol

Specifications

Binding Capacity ∼10 g/L resin
Flow Rate 50 to 200 cm/hr. (Up to 2 bar)
Matrix Agarose-based, aldehyde-activated
Content And Storage Store at 2°C to 8°C.
Particle Size 65 ±10 μm
Form Liquid Suspension
Product Line CaptureSelect
Quantity 5 mL
Type Affinity Matrix
Ligand Type Bovine IgA affinity ligand
Stationary Phase Anti Bovine IgA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.