missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ Biotin Anti-HGH Conjugate

Product Code. 18426798 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
500 μg
Unit Size:
100µg
500µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18426798 100 μg 100µg
18496538 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18426798 Supplier Thermo Scientific™ Supplier No. 7103160100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ CaptureSelect™ Biotin Anti-hGH Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) with high affinity and selectivity for both recombinant and natural sourced human growth hormone (hGH).

CaptureSelect Biotin Anti-hGH Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) with high affinity and selectivity for both recombinant and natural sourced human growth hormone (hGH). The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.

The CaptureSelect Biotin Anti-hGH Conjugate allows you to:

  • Detect, quantitate, and characterize recombinant and natural sourced hGH

Applications for CaptureSelect Biotin Anti-hGH Conjugate include ELISA, Western blot, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5°C to -30°C).
Form Liquid
Product Line CaptureSelect
Quantity 100 μg
Type Ligand Conjugates
Label or Dye Biotin & Other Haptens
Ligand Type Antibody
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.