Learn More
Thermo Scientific™ CaptureSelect™ Biotin Anti-Free LC-kappa (Human) Conjugate
Shop All Thermo Scientific ProductsDescription
CaptureSelect Biotin Anti-Free LC-kappa (Hu) Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that binds highly selectively to free human kappa light chains and dimers thereof. The functionality is based on recognition of an epitope on the human CL-kappa domain that is masked in intact antibodies and Fab fragments. The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.
The CaptureSelect Biotin Anti-Free LC-kappa (Hu) Conjugate allows you to:
- Detect, quantitate, and characterize free human kappa light chains in (crude) antibody preps
- Avoid cross-binding with human kappa light chains present in intact and properly folded antibodies and Fab fragments of any human antibody isotype such as IgG, IgA, IgM, IgD, and IgE
Applications for CaptureSelect Biotin Anti-Free LC-kappa (Hu) Conjugate include ELISA, Western blot, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).
Animal origin statement
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Specifications
Specifications
| Content And Storage | Store in freezer (-5°C to -30°C). |
| Form | Liquid |
| Product Line | CaptureSelect |
| Quantity | 500 μg |
| Type | Conjugate |
| Label or Dye | Biotin & Other Haptens |
| Ligand Type | Antibody |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.