missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ Antithrombin III Affinity Matrix

Product Code. 18456588 Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 mL
10 mL
Unit Size:
10mL
5mL
This item is not returnable. View return policy

Product Code. 18456588

Brand: Thermo Scientific™ 190317010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This is one of a group of Thermo Scientific™ CaptureSelect™ protein purification affinity resins developed for the simple, single-step purification of a variety human plasma and serum proteins.

This is one of a group of CaptureSelect protein purification affinity resins developed for the simple, single-step purification of a variety human plasma and serum proteins. This unique selection of affinity purification resins can purify proteins or other non-antibody biomolecules from either recombinant or native sources with a high degree of affinity and specificity, and are immobilized on a high capacity agarose support.

Specifications

Content And Storage Store at 2°C to 8°C.
Form Liquid Suspension
Product Line CaptureSelect
Quantity 10 mL
Type Bulk Resin
Stationary Phase Anti Antithrombin III
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.