missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ Alpha-1 Antitrypsin Select Leakage ELISA

Product Code. 18476498 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 Assay
10 Assays
Unit Size:
Each
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18476498 1 Assay Each
30275958 10 Assays Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18476498 Supplier Thermo Scientific™ Supplier No. 810287001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ CaptureSelect™ Alpha-1 AntitrypsinSelect Leakage ELISA is designed to detect possible leached ligand of human Alpha-1 antitrypsin (AAT) purified using AAT Select affinity resin (Cytiva).

CaptureSelect Alpha-1 AntitrypsinSelect Leakage ELISA is designed to detect possible leached ligand of human Alpha-1 antitrypsin (AAT) purified using AAT Select affinity resin (Cytiva). Even when covalently attached, small amounts of the Alpha-1 Antitrypsin Select affinity ligand can leach off of the chromatography support and co-elute with the purified product.

This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect Alpha-1 Antitrypsin Select Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.

Specifications

Content And Storage Store at -5°C to -30°C.
Assay ELISA
For Use With (Equipment) Microplate Reader
Product Line CaptureSelect
Quantity 1 Assay
Target Alpha-1 AntitrypsinSelect Affinity Ligand
Type ELISA Kit
Format Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.