missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Capto S™ ImpAct Chromatography Media

Capto S ImpAct is a strong cation exchange chromatography medium (resin) developed for polishing of monoclonal antibodies (MAbs) and other biomolecules.

Brand:  Cytiva 17371702

Code : NEW

Additional Details : CAS Number : 64-17-5 Weight : 0.26700kg


 View more versions of this product

Product Code. 15351642

  • £515.00 / 100mL
Estimated Shipment: 20-05-2024
to see stock.
Add to basket

Description

Description

  • High-flow characteristics of Capto media allows for good pressure-flow properties
  • High binding capacity and resolution, combined with the ability to run at high flow rates and bed heights, increases productivity and flexibility in process design
  • Efficient aggregate removal at high load of monoclonal antibodies
Specifications

Specifications

Capto S ImpAct Chromatography Media
High-flow agarose
50μm
100mL
4 to 12
Ion Exchange Chromatography Media
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Capto S ImpAct, 100 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-18 per cent of the mixture consists of component(s) of unknown acute toxicity.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers