missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Capto™ L Chromatography Medium

Capto L is a BioProcess chromatography medium (resin) for the capture of antibodies and antibody fragments.

Brand:  Cytiva 17-5478-06

Code : GE

Additional Details : CAS Number : 64-17-5 Weight : 0.02500kg


 View more versions of this product

Product Code. 12651206

  • £609.00 / Each
Estimated Shipment: 20-05-2024
to see stock.
Add to basket

Description

Description

Capto L is suitable for the capture of a wide range of antibody fragments such as Fabs, single-chain variable fragments (scFv), and domain antibodies (Dabs).

  • High binding capacity, low ligand leakage, and selectivity for a broad range of antibody fragments reduces process time and amount of medium.
  • Enables a platform approach for purification of antibody fragments.
  • Increased productivity, process economy, and process design flexibility due to rigid matrix.
  • The high binding capacity and selectivity for a broad range of antibody fragments makes Capto L an excellent capture platform for antibody fragments containing kappa light chains.
  • Using Capto L requires minimal optimization and it produces high purity in one separation step.
  • Capto L provides several advantages in manufacturing processes because the high selectivity of affinity capture reduces the number of subsequent polishing steps.

Specifications

Specifications

Capto L chromato graphy media
7.87 in.
<500cm/hr.
2 to 10
Highly Cross-linked Agarose
High binding capacity, low ligand leakage and selectivity for a broad range of Antibody fragments reduces process time and amount of medium
5mL
200mm
Recombinant Protein L (E. coli), mammalian free
85μm
2° to 8°C
Affinity Resin
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • capto L

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P235-Keep cool.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers