Description
- Capto adhere is a multimodal BioProcess resin designed for post-protein A purification of monoclonal antibodies (MAbs) at process scale.
- Composed of a rigid, high-flow agarose matrix and a novel multimodal ligand.
- The strong multimodal ligand gives a different selectivity compared to traditional ion exchangers. Capto adhere can remove key contaminants such as DNA, host cell proteins (HCP), leached protein A, dimers and larger aggregates, and viruses in a single step.
- Together with a Protein A capture step, like MabSelect SuRe, Capto adhere allows the design of a two-step chromatographic process for Mabs.
- For optimal performance of Capto adhere, screening/optimization of loading conditions is recommended, preferably using modern high-throughput process development tools, such as PreDictor 96-well filter plates.
- Capto adhere is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development.
- As member of the BioProcess resins range, Capto adhere meets industrial demands with security of supply and comprehensive technical and regulatory support.
Specifications
Specifications
| Quantity | 100 mL |
| Material | Agarose matrix |
| Ligand Type | Multimodal |
| Matrix | Ion Exchange Chromatography Media |
Safety and Handling
Product Identifier
- capto adhere
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P235-Keep cool.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: ethanol