missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CAPS Monoclonal Antibody (K49040_1E11), Invitrogen™
Mouse Monoclonal Antibody
£416.00 - £734.00
Specifications
| Antigen | CAPS |
|---|---|
| Clone | K49040_1E11 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30207692
|
Invitrogen™
MA555764 |
50 μL |
£416.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30201311
|
Invitrogen™
MA556144 |
200 μL |
£734.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: TTQLTQGPAG RTLGQTQASC PEPRPSMDAV DATMEKLRAQ CLSRGASGIQ GLARFFRQLD RDGSRSLDAD EFRQGLAKLG LVLDQAEAEG VCRKWDRNGS GTLDLEEFLR ALRPPMSQAR EAVIAAAFAK LDRSGDGVVT VDDLRGVYSG RAHPKVRSGE WTEDEVLRRF LDNFDSSEKD GQVTLAEFQD YYSGVSASMN TDEEFVAMMT
CAPS Monoclonal antibody specifically detects CAPS in Human samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| CAPS | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Calcyphosin; calcyphosine; calcyphosine 1; CAPS; CAPS1; thyroid protein p24 | |
| Recombinant human CAPS (immunogen range is 61-270). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| CAPS |
| K49040_1E11 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q13938 | |
| 828 | |
| IgG2b | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title