Learn More
Invitrogen™ CAPS Monoclonal Antibody (K49040_1E11), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556144
Description
Sequence of this protein is as follows: TTQLTQGPAG RTLGQTQASC PEPRPSMDAV DATMEKLRAQ CLSRGASGIQ GLARFFRQLD RDGSRSLDAD EFRQGLAKLG LVLDQAEAEG VCRKWDRNGS GTLDLEEFLR ALRPPMSQAR EAVIAAAFAK LDRSGDGVVT VDDLRGVYSG RAHPKVRSGE WTEDEVLRRF LDNFDSSEKD GQVTLAEFQD YYSGVSASMN TDEEFVAMMT
CAPS Monoclonal antibody specifically detects CAPS in Human samples. It is validated for Immunoprecipitation, Western Blot
Specifications
| CAPS | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q13938 | |
| CAPS | |
| Recombinant human CAPS (immunogen range is 61-270). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunoprecipitation, Western Blot | |
| K49040_1E11 | |
| Unconjugated | |
| CAPS | |
| Calcyphosin; calcyphosine; calcyphosine 1; CAPS; CAPS1; thyroid protein p24 | |
| Mouse | |
| Protein A | |
| RUO | |
| 828 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.