missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAPNS1 Mouse, anti-Human, Unconjugated, Clone: 2B6G6, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | CAPNS1 |
|---|---|
| Clone | 2B6G6 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17271496
|
Proteintech
67838-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17261496
|
Proteintech
67838-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CAPNS1 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human CAPNS1. This antibody recognizes human, rat antigen. The CAPNS1 antibody has been validated for the following applications: WB,ELISA analysis.
Specifications
| CAPNS1 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| Human, Rat | |
| Primary |
| 2B6G6 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| 826 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title