Learn More
CAPNS1 Mouse, anti-Human, Unconjugated, Clone: 2B6G6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67838-1-IG-20UL
This item is not returnable.
View return policy
Description
The CAPNS1 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human CAPNS1. This antibody recognizes human, rat antigen. The CAPNS1 antibody has been validated for the following applications: WB,ELISA analysis.
Specifications
| CAPNS1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat |
| Western Blot, ELISA | |
| 2B6G6 | |
| Unconjugated | |
| 20 μL | |
| 826 | |
| Liquid |