Learn More
SPEX™ Cap-Mat for Square-Well Plate
SPEX™ Cap-Mat is a sealing mat specifically designed for the 2200 96-Well Titer Plate.
Brand: SPEX™ 2201
Code : Z3
Additional Details : Weight : 0.10000kg
Description
- Tightly seals titer plate wells
- Prevents spills and well-to-well contamination
- Made of rugged silicone rubber
- May be sterilized and reused
Specifications
2200 96-Well Titer Plate | |
Silicone Rubber |