missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Canine RAGE Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF4750
This item is not returnable.
View return policy
Description
AGER Polyclonal specifically detects AGER in Canine samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Specifications
| RAGE | |
| Polyclonal | |
| Unconjugated | |
| XP_532093 | |
| MOK | |
| Mouse myeloma cell line NS0-derived recombinant canine RAGE Asp25-Val339 Accession # XP_532093 | |
| 100 μg | |
| Primary | |
| Detects canine RAGE in ELISAs and Western blots. In sandwich immunoassays, approximately 0.5% cross-reactivity with recombinant human RAGE is observed and less than 0.1% cross-reactivity with recombinant mouse RAGE and recombinant rat RAGE is observed. | |
| Canine | |
| IgG |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| advanced glycosylation end product-specific receptor, AGER, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products, SCARJ1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 5891 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction