missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Canine RAGE Antibody, R&D Systems™

Sheep Polyclonal Antibody
£167.00 - £452.00
Specifications
| Antigen | RAGE |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18737497
![]() |
R&D Systems
AF4750-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18061082
|
R&D Systems
AF4750 |
100 μg |
£452.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AGER Polyclonal specifically detects AGER in Canine samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
| RAGE | |
| Western Blot 0.1 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
| Polyclonal | |
| Sheep | |
| Canine | |
| advanced glycosylation end product-specific receptor, AGER, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products, SCARJ1 | |
| MOK | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| XP_532093 | |
| 5891 | |
| Mouse myeloma cell line NS0-derived recombinant canine RAGE Asp25-Val339 Accession # XP_532093 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects canine RAGE in ELISAs and Western blots. In sandwich immunoassays, approximately 0.5% cross-reactivity with recombinant human RAGE is observed and less than 0.1% cross-reactivity with recombinant mouse RAGE and recombinant rat RAGE is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title