missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Canine HGF Antibody, R&D Systems™

Sheep Polyclonal Antibody
£157.00 - £432.00
Specifications
| Antigen | HGF |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18715577
![]() |
R&D Systems
AF3386-SP |
25 μg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18041712
|
R&D Systems
AF3386 |
100 μg |
£432.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HGF Polyclonal specifically detects HGF in Canine samples. It is validated for Western Blot.Specifications
| HGF | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Sheep | |
| Canine | |
| deafness, autosomal recessive 39, DFNB39, EC 3.4.21, EC 3.4.21.7, fibroblast-derived tumor cytotoxic factor, F-TCF, hepatocyte growth factor (hepapoietin A; scatter factor), Hepatopoeitin-A, Hepatopoietin A, HGFB, HPTA, HPTAhepatocyte growth factor, lung fibroblast-derived mitogen, Scatter factor, SF, SFhepatopoeitin-A | |
| HGF | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q867B7 | |
| 3082 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant canine HGF Arg28-Arg494 (Lys266Gln, Trp321Arg, Leu458Pro) ( alpha chain) & Val495-Ser730 (Ile693Thr) ( beta chain) Accession # Q867B7 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects canine HGF in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant human (rh) HGF is observed, approximately 15% cross-reactivity with recombinant mouse HGF is observed, and less than 3% cross-reactivity with rhMSP is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title