missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Canine HGF Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF3386
This item is not returnable.
View return policy
Description
HGF Polyclonal specifically detects HGF in Canine samples. It is validated for Western Blot.
Specifications
| HGF | |
| Polyclonal | |
| Unconjugated | |
| Q867B7 | |
| HGF | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant canine HGF Arg28-Arg494 (Lys266Gln, Trp321Arg, Leu458Pro) ( alpha chain) & Val495-Ser730 (Ile693Thr) ( beta chain) Accession # Q867B7 | |
| 100 μg | |
| Primary | |
| Detects canine HGF in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant human (rh) HGF is observed, approximately 15% cross-reactivity with recombinant mouse HGF is observed, and less than 3% cross-reactivity with rhMSP is observed. | |
| Canine | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| deafness, autosomal recessive 39, DFNB39, EC 3.4.21, EC 3.4.21.7, fibroblast-derived tumor cytotoxic factor, F-TCF, hepatocyte growth factor (hepapoietin A; scatter factor), Hepatopoeitin-A, Hepatopoietin A, HGFB, HPTA, HPTAhepatocyte growth factor, lung fibroblast-derived mitogen, Scatter factor, SF, SFhepatopoeitin-A | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 3082 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction