missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Campylobacter C.A.T. Agar

Product Code. 12927178
This item is not returnable. View return policy
Product Code. 12927178 Supplier Thermo Scientific™ Supplier No. PO0839A

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Isolate thermophilic Campylobacter species and experience improved recovery of Campylobacter upsaliensis from feces with this selective medium.

Isolate thermophilic Campylobacter species and experience improved recovery of Camplyobacter upsaliensis from feces with ready-to-use Thermo Scientific™ Oxoid™ Campylobacter C.A.T. Agar. Campylobacter C.A.T. Agar is a modification of the formulation described by Aspinall1, in which blood has been replaced with charcoal, ferrous sulphate and sodium pyruvate. Comparative studies with blood-free and blood-containing selective media for campylobacters have shown that the blood-free, charcoal-containing media are superior to those containing blood, increasing the aerotolerance of these organisms.2,3

Use Campylobacter C.A.T. Agar for the isolation of thermophilic Campylobacter species.

  • Suitable for isolation of thermophilic Campylobacter.
  • Improved recovery of Campylobacter upsaliensis.
  • Ready-to-use convenience of prepared media.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Content And Storage 2°C to 10°C
Format 90mm Monoplate
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight 19.5g ± 1.0g
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.