missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX cAMP Hunter™ Liraglutide Bioassay Kit
Description
The cAMP Hunter Liraglutide Bioassay kit provides an easy-to-use cell based assay to determine Liraglutide potency and detect neutralizing antibodies. This bioassay assesses Liraglutide based activation of GLP-1R GPCR activity via detection of levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wildtype GLP-1R , a naturally Gs coupled GPCR. Upon activation of GLP-1R , the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates.
Specifications
Specifications
| Assay Entry | Intracellular cAMP level |
| For Use With (Application) | Assay-Ready Bioassay Kits |
| Quantity | 1,000 dp (10 x 96-well) |
| Target Entry | GLP-1R |
| Cell Line | CHO-K1 |
| Target | GLP-1R |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?