missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX cAMP Hunter™ eXpress GRM6 CHO-K1 GPCR Assay
Click to view available options
Quantity:
200 dp (2 x 96-well)
Unit Size:
1 set
Description
The cAMP Hunter™ GRM6 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. L-glutamate) based activation of GRM6 GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wildtype GRM6 , a naturally Gi/Go coupled GPCR. Upon activation of GRM6 , the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Specifications
Specifications
| Assay Entry | CHO-K1 |
| For Use With (Application) | Assay-Ready eXpress Kits |
| Primary Agonist | L-glutamate |
| Quantity | 200 dp (2 x 96-well) |
| Target Entry | GRM6 |
| Cell Line | CHO-K1 |
| Target | GRM6 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction