missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CaMKIV Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£159.00
Specifications
| Antigen | CaMKIV |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5 μg/mL, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Frozen 1 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
CaMKIV Polyclonal specifically detects CaMKIV in Human, Mouse, Rat, Canine, Plant, Primate, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
| CaMKIV | |
| Western Blot 0.5 μg/mL, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Frozen 1 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Cellular Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission, Protein Kinase | |
| PBS with 0.05% Sodium Azide | |
| 814 | |
| Synthetic Peptide: V(127) E K G Y Y S E R D A A D A V K Q(143) C | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| brain Ca(2+)-calmodulin-dependent protein kinase type IV, brain Ca++-calmodulin-dependent protein kinase type IV, calcium/calmodulin-dependent protein kinase IV, calcium/calmodulin-dependent protein kinase type IV, calcium/calmodulin-dependent protein kinase type IV catalytic chain, CAM kinase- GR, CAM kinase IV, CaM kinase-GR, caMK, CaMK IV, CaMK-GR, EC 2.7.11, EC 2.7.11.17, IV, MGC36771 | |
| CAMK4 | |
| IgG | |
| Affinity Purified | |
| NB120-3557 detects CaM kinase IV (CaMKIV) from human, mouse, rat, canine, primate, arabidopsis and Xenopus Oocyte samples. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title