missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CaMKIV Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB120-3557-0.025mg
This item is not returnable.
View return policy
Description
CaMKIV Polyclonal specifically detects CaMKIV in Human, Mouse, Rat, Canine, Plant, Primate, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| CaMKIV | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| CAMK4 | |
| Synthetic Peptide: V(127) E K G Y Y S E R D A A D A V K Q(143) C | |
| 0.025 mg | |
| Cellular Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission, Protein Kinase | |
| 814 | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 0.5 μg/mL, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Frozen 1 μg/mL | |
| brain Ca(2+)-calmodulin-dependent protein kinase type IV, brain Ca++-calmodulin-dependent protein kinase type IV, calcium/calmodulin-dependent protein kinase IV, calcium/calmodulin-dependent protein kinase type IV, calcium/calmodulin-dependent protein kinase type IV catalytic chain, CAM kinase- GR, CAM kinase IV, CaM kinase-GR, caMK, CaMK IV, CaMK-GR, EC 2.7.11, EC 2.7.11.17, IV, MGC36771 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| NB120-3557 detects CaM kinase IV (CaMKIV) from human, mouse, rat, canine, primate, arabidopsis and Xenopus Oocyte samples. | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction