missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CaM Kinase I Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£155.00
Specifications
| Antigen | CaM Kinase I |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 2 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18502444
|
Novus Biologicals
NB600-1163-0.025mg |
0.025 mg | |||||||
Description
CaM Kinase I Polyclonal specifically detects CaM Kinase I in Mouse, Rat samples. It is validated for Western Blot.Specifications
| CaM Kinase I | |
| Western Blot 2 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Protein Kinase | |
| PBS and 1 mg/ml BSA with 0.05% Sodium Azide | |
| 8536 | |
| Synthetic Peptide: A(1) V E G P R W K Q A E D I R D I Y D(18) | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Mouse | |
| calcium/calmodulin-dependent protein kinase I, calcium/calmodulin-dependent protein kinase type 1, CaM kinase I, CaM kinase I alpha, caM-KI, CaMKI, CaMKI-alpha, EC 2.7.11, EC 2.7.11.17, MGC120317, MGC120318 | |
| CAMK1 | |
| IgG | |
| Affinity Purified | |
| Detects CaMKI from rat brain extract. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title