missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CaM Kinase I Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-1163-0.025mg
This item is not returnable.
View return policy
Description
CaM Kinase I Polyclonal specifically detects CaM Kinase I in Mouse, Rat samples. It is validated for Western Blot.
Specifications
| CaM Kinase I | |
| Polyclonal | |
| Unconjugated | |
| PBS and 1 mg/ml BSA with 0.05% Sodium Azide | |
| CAMK1 | |
| Synthetic Peptide: A(1) V E G P R W K Q A E D I R D I Y D(18) | |
| 0.025 mg | |
| Protein Kinase | |
| 8536 | |
| Mouse | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 2 μg/mL | |
| calcium/calmodulin-dependent protein kinase I, calcium/calmodulin-dependent protein kinase type 1, CaM kinase I, CaM kinase I alpha, caM-KI, CaMKI, CaMKI-alpha, EC 2.7.11, EC 2.7.11.17, MGC120317, MGC120318 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Detects CaMKI from rat brain extract. | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction