missing translation for 'onlineSavingsMsg'
Learn More

Calponin Rabbit anti-Human, Mouse, Rat, CoraLite 594, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £375.00

Specifications

Antigen Calponin
Concentration 1000 μg/mL
Applications Immunofluorescence
Classification Polyclonal
Conjugate CoraLite 594
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17791379
View Documents
Proteintech
CL594-13938-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17781379
View Documents
Proteintech
CL594-13938-100UL
100 μL
£375.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Calponin Polyclonal antibody specifically detects Calponin in Human, Mouse, Rat samples. It is validated for IF
Specifications

Specifications

Calponin
Immunofluorescence
CoraLite 594
Rabbit
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
IgG
Antigen affinity purification
Antibody
1000 μg/mL
Polyclonal
Liquid
Human, Mouse, Rat
1264
Primary
Store at -20°C
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.