missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calponin Rabbit anti-Human, Mouse, Rat, CoraLite 594, Polyclonal, Proteintech
Shop All Proteintech ProductsDescription
Calponin Polyclonal antibody specifically detects Calponin in Human, Mouse, Rat samples. It is validated for IF
Specifications
Specifications
| Antigen | Calponin |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 1000 μg/mL |
| Conjugate | CoraLite 594 |
| Formulation | PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. |
| Gene Accession No. | BC016774 |
| Host Species | Rabbit |
| Purification Method | Antigen affinity purification |
| Quantity | 20 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?