missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Calponin 2 Monoclonal Antibody (C8), Invitrogen™
Mouse Monoclonal Antibody
£161.00 - £369.00
Specifications
| Antigen | Calponin 2 |
|---|---|
| Clone | C8 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17929692
|
Invitrogen™
MA551028 |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17989202
|
Invitrogen™
MA551027 |
200 μL |
£369.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene, which can bind actin, calmodulin, troponin C, and tropomyosin, may function in the structural organization of actin filaments. The encoded protein could play a role in smooth muscle contraction and cell adhesion. Two transcript variants encoding different isoforms have been found for this gene.Specifications
| Calponin 2 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| AA408047; AI324678; Calpo2; calponin 2; calponin H2, smooth muscle; Calponin-2; CLP2; CNN2; h2-calponin; LOW QUALITY PROTEIN: calponin-2; neutral calponin | |
| Cnn2 | |
| IgG1 κ | |
| Protein A/G | |
| Cnn2 |
| C8 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Pig | |
| Q08094, Q99439 | |
| 1265, 397565 | |
| Recombinant Human CNN2 protein, Met1-Gln121+Ala131-Tyr269 (Accession Q99439). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title