missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Calponin 2 Monoclonal Antibody (C8), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA551027
This item is not returnable.
View return policy
Description
The protein encoded by this gene, which can bind actin, calmodulin, troponin C, and tropomyosin, may function in the structural organization of actin filaments. The encoded protein could play a role in smooth muscle contraction and cell adhesion. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| Calponin 2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| Q08094, Q99439 | |
| Cnn2 | |
| Recombinant Human CNN2 protein, Met1-Gln121+Ala131-Tyr269 (Accession Q99439). | |
| 200 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG1 κ |
| Western Blot | |
| C8 | |
| Unconjugated | |
| Cnn2 | |
| AA408047; AI324678; Calpo2; calponin 2; calponin H2, smooth muscle; Calponin-2; CLP2; CNN2; h2-calponin; LOW QUALITY PROTEIN: calponin-2; neutral calponin | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 1265, 397565 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction