Learn More
Thermo Scientific Chemicals Calpain Inhibitor I, 95+%
Description
It inhibits apoptosis in thymocytes and metamyelocytes. It also inhibits cell cycle progression at the G1/S border and metaphase/anaphase in CHO cells by inhibiting cyclin B degradation. ALLN inhibits the proteolysis of IκBα and IκBβ by the ubiquitin-proteasome complex. ALLN also prevents nitric oxide production by activated macrophages by interfering with transcription of the inducible nitric oxide synthase gene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 5 mg |
| Description | White |
| Content And Storage | -30°C to -10°C |
| For Use With (Application) | An inhibitor of both Calpain I and II |
| CAS | 110044-82-1 |
| Molecular Formula | C20H37N3O4 |
| Formula Weight | 383.53 |
| Synonym | Ac-LLnL-CHO; ALLN |
| Form | Powder |
| Assay | 95+% |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.