missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calpain 3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £359.00
Specifications
| Antigen | Calpain 3 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18640761
|
Novus Biologicals
NBP2-92075-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18618401
|
Novus Biologicals
NBP2-92075-0.1ml |
0.1 mL |
£359.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calpain 3 Polyclonal antibody specifically detects Calpain 3 in Human samples. It is validated for Western BlotSpecifications
| Calpain 3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Neuroscience, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 825 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| calpain 3, (p94), Calpain L3, Calpain p94, calpain, large polypeptide L3, calpain-3, CANP 3, CANP3MGC4403, CANPL3, EC 3.4.22, EC 3.4.22.54, LGMD2, LGMD2A, MGC10767, MGC11121, MGC14344, Muscle-specific calcium-activated neutral protease 3, muscle-specific calcium-activated neutral protease 3 large subunit, NCL1, nCL-1large [catalytic] subunit, New calpain 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human CAPN3 (NP_775111.1). MHGNKQHLQKDFFLYNASKARSKTYINMREVSQRFRLPPSEYVIVPSTYEPHQEGEFILRVFSEKRNLSEEVENTISVDRPVKKKKTKPIIFVSDRANSNKELGVDQESEEGKGKTSPDKQKQSPQPQPGSSDQESEEQQQFRNIFKQIA | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts