missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calpain 3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92075-0.1ml
This item is not returnable.
View return policy
Description
Calpain 3 Polyclonal antibody specifically detects Calpain 3 in Human samples. It is validated for Western Blot
Specifications
| Calpain 3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| calpain 3, (p94), Calpain L3, Calpain p94, calpain, large polypeptide L3, calpain-3, CANP 3, CANP3MGC4403, CANPL3, EC 3.4.22, EC 3.4.22.54, LGMD2, LGMD2A, MGC10767, MGC11121, MGC14344, Muscle-specific calcium-activated neutral protease 3, muscle-specific calcium-activated neutral protease 3 large subunit, NCL1, nCL-1large [catalytic] subunit, New calpain 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human CAPN3 (NP_775111.1). MHGNKQHLQKDFFLYNASKARSKTYINMREVSQRFRLPPSEYVIVPSTYEPHQEGEFILRVFSEKRNLSEEVENTISVDRPVKKKKTKPIIFVSDRANSNKELGVDQESEEGKGKTSPDKQKQSPQPQPGSSDQESEEQQQFRNIFKQIA | |
| 0.1 mL | |
| Cell Biology, Neuroscience, Signal Transduction | |
| 825 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction