missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calnexin, Mouse anti-Human, Clone: CANX/1543, Novus Biologicals™
Mouse Monoclonal Antibody
£221.00 - £396.00
Specifications
| Antigen | Calnexin |
|---|---|
| Clone | CANX/1543 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1-2 ug/ml, ELISA 2-4 ug/ml, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18229877
|
Novus Biologicals
NBP2-79701-20UG |
20 μg |
£221.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18237307
|
Novus Biologicals
NBP2-79701-100UG |
100 μg |
£396.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calnexin Monoclonal specifically detects Calnexin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifikationer
| Calnexin | |
| 0.2 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| ER Markers, Neuroscience, Phospho Specific, Sensory Systems, Unfolded Protein Response, Vision | |
| 10 mM PBS with 0.05% BSA | |
| calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90 | |
| CANX | |
| IgG1 κ | |
| Protein A or G purified | |
| This antibody recognizes a protein of 90kDa, which is identified as Calnexin. |
| CANX/1543 | |
| Western Blot 1-2 ug/ml, ELISA 2-4 ug/ml, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P27824 | |
| 821 | |
| Recombinant N-terminal fragment of human Calnexin protein (around aa 1-300) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel