missing translation for 'onlineSavingsMsg'
Learn More

Calnexin, Mouse anti-Human, Clone: CANX/1543, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-79701-100UG

 View more versions of this product

Product Code. 18237307

  • £396.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Calnexin Monoclonal specifically detects Calnexin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Calnexin
Monoclonal
0.2 mg/mL
Western Blot 1-2 ug/ml, ELISA 2-4 ug/ml, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
P27824
CANX
Recombinant N-terminal fragment of human Calnexin protein (around aa 1-300) (exact sequence is proprietary)
100 μg
ER Markers, Neuroscience, Phospho Specific, Sensory Systems, Unfolded Protein Response, Vision
821
Human
Purified
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array
CANX/1543
Unconjugated
10 mM PBS with 0.05% BSA
calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90
Mouse
Protein A or G purified
RUO
Primary
This antibody recognizes a protein of 90kDa, which is identified as Calnexin.
Store at 4°C.
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.