missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calmodulin Antibody (C16T), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-29791
This item is not returnable.
View return policy
Description
Calmodulin Monoclonal specifically detects Calmodulin in Vertebrate samples. It is validated for Western Blot.
Specifications
| Calmodulin | |
| Monoclonal | |
| Unconjugated | |
| CALM, CALM2, CALM3, CALML2, calmodulin, calmodulin 1 (phosphorylase kinase, delta), CaM, CAM1, CAM2, CAM3, CAMB, CAMC, CAMI, CAMIII, DD132, PHKDCAM, phosphorylase kinase, delta subunit | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Recombinant proteins containing the calmodulin binding protein epitope tag. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| C16T | |
| Western Blot 1:100-1:2000 | |
| CALM1 | |
| KLH-conjugated, synthetic peptide corresponding to the Calmodulin Binding Protein (CBP) epitope tag found in several E. coli expression vectors. | |
| 0.1 mL | |
| Cell Cycle and Replication, Neuroscience, Neurotransmission, Signal Transduction | |
| 801 | |
| Other | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction