missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calmodulin Antibody (C16T), Novus Biologicals™
Rabbit Monoclonal Antibody
£145.00 - £426.00
Specifications
| Antigen | Calmodulin |
|---|---|
| Clone | C16T |
| Dilution | Western Blot 1:100-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18162544
|
Novus Biologicals
NBP2-29791-0.025ML |
0.025 mL |
£145.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18185912
|
Novus Biologicals
NBP2-29791 |
0.1 mL |
£426.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calmodulin Monoclonal specifically detects Calmodulin in Vertebrate samples. It is validated for Western Blot.Specifications
| Calmodulin | |
| Western Blot 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Other | |
| 801 | |
| KLH-conjugated, synthetic peptide corresponding to the Calmodulin Binding Protein (CBP) epitope tag found in several E. coli expression vectors. | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| C16T | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, Neuroscience, Neurotransmission, Signal Transduction | |
| CALM, CALM2, CALM3, CALML2, calmodulin, calmodulin 1 (phosphorylase kinase, delta), CaM, CAM1, CAM2, CAM3, CAMB, CAMC, CAMI, CAMIII, DD132, PHKDCAM, phosphorylase kinase, delta subunit | |
| CALM1 | |
| IgG | |
| Protein A purified | |
| Recombinant proteins containing the calmodulin binding protein epitope tag. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title