missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CALML5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£169.00 - £255.00
Specifications
| Antigen | CALML5 |
|---|---|
| Dilution | ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18621997
|
Novus Biologicals
NBP2-99872-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18616807
|
Novus Biologicals
NBP2-99872-50ul |
50 μg |
£169.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CALML5 Polyclonal antibody specifically detects CALML5 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
| CALML5 | |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cell Cycle and Replication, Neurodegeneration, Neuroscience, Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 51806 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| calmodulin-like 5, calmodulin-like protein 5, CLSPCalmodulin-like skin protein | |
| Produced in rabbits immunized with purified, recombinant Human CALML5 (Accession#: AAH39172.1; Met 1-Glu 146) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title