missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CALML5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99872-50ul
This item is not returnable.
View return policy
Description
CALML5 Polyclonal antibody specifically detects CALML5 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
| CALML5 | |
| Polyclonal | |
| ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| calmodulin-like 5, calmodulin-like protein 5, CLSPCalmodulin-like skin protein | |
| Produced in rabbits immunized with purified, recombinant Human CALML5 (Accession#: AAH39172.1; Met 1-Glu 146) | |
| 50 μg | |
| Cell Biology, Cell Cycle and Replication, Neurodegeneration, Neuroscience, Signal Transduction | |
| 51806 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering