missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Calmidazolium Chloride

Product Code. 15455539
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15455539 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15455539 Supplier Thermo Scientific Chemicals Supplier No. J63270.MA
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 57265-65-3 | C31H23Cl7N2O, C31H24Cl7N2O | 688.70 g/mol

Calmidazolium chloride is a small molecule CaM antagonist and calcium channel protein inhibitor. Calmidazolium chloride demonstrates noncompetitive inhibition of platelet and sarcoplasmic reticulum Ca2+-ATPase activity and shows inhibition of Ca2+ transport activity in platelet microsomes. In HeLa cells, calmidazolium chloride stimulates a mechanism of cellular Ca2+ influx distinct from store-operated Ca2+ entry and unaccompanied by Ca2+ store depletion, mobilizing stores and evoking a Ca2+ signal through activation of phospholipase C. Calmidazolium chloride is described to inhibit constitutive nitric oxide synthase (NOS) activity without affecting inducible NOS in rat aorta. The dependence upon calmodulin of neuronal NOS (nNOS) in neuroblastoma N1E-115 cells was probed with calmidazolium chloride, where it was demonstrated to increase cGMP levels indirectly indicating an increase in NO output by nNOS. Calmidazolium chloride is also described to inhibit the activity of calmodulin N-methyltransferase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Description White
Format Solid
For Use With (Application) Inhibits the calcium-calmodulin dependent phosphodiesterase
CAS 57265-65-3
Molecular Formula C31H23Cl7N2O
Formula Weight 687.71
Form Solid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.