missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CALM Recombinant Rabbit Monoclonal Antibody (24GB1620)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557603
This item is not returnable.
View return policy
Description
CALM Recombinant Monoclonal Antibody for Western Blot, Flow
PICALM, also known as the clathrin assembly lymphoid myeloid leukemia (CALM) protein, co-localizes with clathrin at clathrin-coated areas of the plasma membrane and in the Golgi area, and is thought to be an important component of the coated pit internalization machinery. Fusion proteins involving PICALM have been implicated in acute myeloid and T-cell acute lymphoblastic leukemia. PICALM is also expressed in neurons and is present at the synapse where it facilitates the endocytosis of the synaptic vesicle protein VAMP2. Recent studies have identified variants of PICALM associated with Alzheimer's disease and have suggested this protein may play a role in the endocytosis and recycling of the amyloid protein precursor (APP).
Specifications
| CALM | |
| Recombinant Monoclonal | |
| 0.7 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q13492 | |
| Picalm | |
| A synthesized peptide derived from human PICALM (aa 550-600). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1620 | |
| Unconjugated | |
| Picalm | |
| CALM; Clathrin assembly lymphoid myeloid leukemia; clathrin assembly lymphoid myeloid leukemia protein; clathrin-assembly lymphoid leukemia protein; clathrin-assembly lymphoid myeloid leukemia protein; CLTH; fit1; fit-1; LAP; mKIAA4114; PAP180; phosphatidylinositol binding clathrin assembly protein; phosphatidylinositol-binding clathrin assembly protein; PICALM; rCALM | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 8301 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction