missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caldesmon/CALD1 Mouse anti-Human, Clone: rCALD1/820, Novus Biologicals™
Mouse Monoclonal Antibody
£387.00
Specifications
| Antigen | Caldesmon/CALD1 |
|---|---|
| Clone | rCALD1/820 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, Immunohistochemistry-Paraffin 0.25-0.5 μg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
Description
Caldesmon/CALD1 Monoclonal specifically detects Caldesmon/CALD1 in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.Specifications
| Caldesmon/CALD1 | |
| 0.2 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Cellular Markers, Phospho Specific | |
| 10 mM PBS with 0.05% BSA | |
| 800 | |
| IgG1 κ | |
| Protein A or G purified |
| rCALD1/820 | |
| Western Blot 1 to 2 μg/mL, Immunohistochemistry-Paraffin 0.25-0.5 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CAD, caldesmon, caldesmon 1, CDMH-CAD, HCAD, LCAD, L-CAD, MGC21352, NAG22 | |
| Recombinant full-length human Caldesmon/CALD1 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title