missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caldesmon/CALD1 Mouse anti-Human, Clone: rCALD1/820, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07367-100ug
This item is not returnable.
View return policy
Description
Caldesmon/CALD1 Monoclonal specifically detects Caldesmon/CALD1 in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
Specifications
| Caldesmon/CALD1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1 to 2 μg/mL, Immunohistochemistry-Paraffin 0.25-0.5 μg/mL | |
| CAD, caldesmon, caldesmon 1, CDMH-CAD, HCAD, LCAD, L-CAD, MGC21352, NAG22 | |
| Recombinant full-length human Caldesmon/CALD1 protein | |
| 100 μg | |
| Cell Biology, Cellular Markers, Phospho Specific | |
| 800 | |
| Store at 4°C. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| rCALD1/820 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction