missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Calcium Sensor Dye eFluor™ 514
Membrane-permeable dye used to monitor changes in intracellular free calcium concentrations in the cell. Once eFluor™ 514 Calcium Sensor Dye enters the cell, cellular esterases cleave the AM group yielding a membrane-impermeable dye fluorescing at ≈520nm.
£209.00 - £273.00
Specifications
Detection Method | Fluorescent |
---|---|
For Use With (Application) | Calcium Assay |
For Use With (Equipment) | Fluorescence Microscope, Flow Cytometer |
Product Type | Dye |
Emission | 490⁄514 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15530607
|
Invitrogen™
65-0859-70 |
10 x 50μg |
£209.00
Pack of 10 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15520607
|
Invitrogen™
65-0859-39 |
1mg |
£273.00
1mg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Calcium binding affinity: Kd = 232 nM
- Molecular Weight: ≈1100 d
- Peak Excitation: 490nm
- Peak Emission: 514nm
- Reconstitute in high-quality, freshly opened DMSO
- Recommended concentration: 2 to 5 mM
- Once reconstituted, protect from light and store dessicated at −20°C; avoid freeze-thawing
Specifications
Fluorescent | |
Fluorescence Microscope, Flow Cytometer | |
490⁄514 |
Calcium Assay | |
Dye | |
eBioscience™ |
For Research Use Only. Not for use in diagnostic procedures.