missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Calcium Sensor Dye eFluor™ 514
Click to view available options
Quantity:
10 x 50μg
1mg
Unit Size:
1mg
Pack of 10
Description
- Calcium binding affinity: Kd = 232 nM
- Molecular Weight: ≈1100 d
- Peak Excitation: 490nm
- Peak Emission: 514nm
- Reconstitute in high-quality, freshly opened DMSO
- Recommended concentration: 2 to 5 mM
- Once reconstituted, protect from light and store dessicated at −20°C; avoid freeze-thawing
Specifications
Specifications
| Detection Method | Fluorescence |
| For Use With (Application) | Calcium Assay |
| For Use With (Equipment) | Fluorescence Microscope, Flow Cytometer |
| Product Type | Dye |
| Emission | 490/514 |
| Product Line | eBioscience |
| Quantity | 10 x 50μg |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction