missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Calcineurin Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30789-25ug
This item is not returnable.
View return policy
Description
Calcineurin Polyclonal antibody specifically detects Calcineurin in Bovine,Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry
Specifications
| Calcineurin | |
| Polyclonal | |
| Phosphate buffered saline | |
| The immunogen used for the anti-Calcineurin was a purified peptide conjugated to KLH. The sequence of the peptide is NH2-Arg-Met-Pro-Pro-Arg-Arg-Asp-Ala-Met-Pro-Ser-Asp-Ala-C00H (a.a. 268-280) corresponding to a 60kDa protein | |
| 25 μg | |
| Primary | |
| Bovine, Human, Mouse, Rat | |
| Purified |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Sheep | |
| Ammonium sulfate precipitation | |
| RUO | |
| 5530 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction