missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Calcineurin Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
£162.00 - £386.00
Specifications
| Antigen | Calcineurin |
|---|---|
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066923
|
Novus Biologicals™
NBP3-30789-25ug |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066772
|
Novus Biologicals™
NBP3-30789-100ug |
100 μg |
£386.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calcineurin Polyclonal antibody specifically detects Calcineurin in Bovine,Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,ImmunocytochemistrySpecifications
| Calcineurin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Phosphate buffered saline | |
| The immunogen used for the anti-Calcineurin was a purified peptide conjugated to KLH. The sequence of the peptide is NH2-Arg-Met-Pro-Pro-Arg-Arg-Asp-Ala-Met-Pro-Ser-Asp-Ala-C00H (a.a. 268-280) corresponding to a 60kDa protein | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Sheep | |
| Bovine, Human, Mouse, Rat | |
| 5530 | |
| IgG | |
| Ammonium sulfate precipitation |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title